missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NT5C2 Protein
A cDNA sequence encoding the NT5C2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12888-5ug
Additional Details : Weight : 0.01000kg
Specifications
NT5C2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSTSWSDRLQ NAADMPANMD KHALKKYRRE AYHRVFVNRS LAMEKIKCFG FDMDYTLAVY KSPEYESLGF ELTVERLVSI GYPQELLSFA YDSTFPTRGL VFDTLYGNLL KVDAYGNLLV CAHGFNFIRG PETREQYPNK FIQRDDTERF YILNTLFNLP ETYLLACLVD FFTNCPRYTS CETGFKDGDL FMSYRSMFQD VRDAVDWVHY KGSLKEKTVE NLEKYVVKDG KLPLLLSRMK EVGKVFLATN SDYKYTDKIM TYLFDFPHGP KPGSSHRPWQ SYFDLILVDA RKPLFFGEGT VLRQVDTKTG KLKIGTYTGP LQHGIVYSGG SSDTICDLLG AKGKDILYIG DHIFGDILKS KKRQGWRTFL VIPELAQELH VWTDKSSLFE ELQSLDIFLA ELYKHLDSSS NERPDISSIQ RRIKKVTHDM DMCYGMMGSL FRSGSRQTLF ASQVMRYADL YAASFINLLY YPFSYLFRAA HVLMPHESTV EHTHVDINEM ESPLATRNRT SVDFKDTDYK RHQLTRSISE IKPPNLFPLA PQEITHCHDE DDDEEEEEEE E | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NT5C2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.1M NaCl, 1mM DTT and 0.1mM PMSF. |