missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Orofacial cleft 1 candidate gene 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Orofacial cleft 1 candidate gene 1 protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7807-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
Orofacial cleft 1 candidate gene 1 Protein | |
Research Use Only | |
OFCC1 | |
Recombinant Protein | |
E. coli | |
REKFQQKALK QTKQKKSKSA EFLMVKEDRE ATEGTGNPAF NMSSPDLSAC QTAEKKVIRH DMPDRTLAAH QQKFRLPASA EPKGNEYGRN YFDPLMDEEI NPRQCATEVS REDDDRIFYN RLTKLFDESR QGEPQDESGR EETLNSEAPG SSNKSHEIHK EASEATTAHL EEFQRSQKTI ILLGSSPLEQ EIRSTSLHCM EDEMSHPWIL LLKVTAVIRS RRYYREQRF | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |