missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human OX40 Ligand/TNFSF4 Protein

Product Code. 18725955
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18725955

Brand: R&D Systems™ 1054OX025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to induce IL-8 secretion in HT1080 human fibrosarcoma cells transfected with human OX40/TNFRSF4. Muller, N. et al. (2008) FEBS J. 275:2296. The ED50 for this effect is typically 1-5 ng/mL. Measured by its binding ability in a functional ELISA. When Recombinant Human OX40/TNFRSF4 Fc Chimera (Catalog # 3388-OX ) is immobilized at 10 ng/mL, the concentration of Recombinant Human OX40 Ligand/TNFSF4 that produces 50% optimal binding response is found to be approximately 0.5-3 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6FGS4
For Use With (Application) Bioactivity, Binding Activity
Formulation Supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 7292
Molecular Weight (g/mol) M.W.-observed: 28-34 kDa, reducing conditions, M.W.-theroretical: 18 kDa
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived human OX40 Ligand/TNFSF4 protein MHHHHHHHHHH GGGSGGGSGGGS IEGR Human OX40 Ligand (Gln51-Leu183) (N-terminus) Accession # Q6FGS4 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, O x 40 antigen ligand, O x 40 ligand, O x 40 L, OX-40 L, O x 4 OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34 kD), tax-transcriptionally activated glycoprotein 1, 34 kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Gene Symbol TNFSF4
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein OX40 Ligand/TNFSF4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.