missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Oxysterols receptor LXR-alpha Protein
A DNA sequence encoding the Homo sapiens (Human) Oxysterols receptor LXR-alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7567-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001123573.1 | |
Oxysterols receptor LXR-alpha Protein | |
Research Use Only | |
NR1H3 | |
Recombinant Protein | |
E. coli | |
MPHSAGGTAG VGLEAAEPTA LLTRAEPPSE PTEIRPQKRK KGPAPKMLGN ELCSVCGDKA SGFHYNVLSC EGCKGFFRRS VIKGAHYICH SGGHCPMDTY MRRKCQECRL RKCRQAGMRE ECVLSEEQIR LKKLKRQEEE QAHATSLPPR ASSPPQILPQ LSPEQLGMIE KLVAAQQQCN RRSFSDRLRV TPWPMAPDPH SREARQQRFA HFTELAIVSV QEIVDFAKQL PGFLQLSRED QIALLKTSAI EVMLLETSRR YNPGSESITF LKDFSYNRED FAKAGLQVEF INPIFEFSRA MNELQLNDAE FALLIAISIF SADRPNVQDQ LQVERLQHTY VEALHAYVSI HHPHDRLMFP RMLMKLVSLR TLSSVHSEQV FALRLQDKKL PPLLSEIWDV HE | |
Greater than 90% as determined by SDS-PAGE. |
10062 | |
200 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |