missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Oxysterols receptor LXR-alpha Protein

Product Code. 16089440
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16089440

Brand: enQuireBio™ QP7567ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Oxysterols receptor LXR-alpha, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001123573.1
Gene ID (Entrez) 10062
Name Oxysterols receptor LXR-alpha Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol NR1H3
Protein Length Full Length of Isoform 3
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MPHSAGGTAG VGLEAAEPTA LLTRAEPPSE PTEIRPQKRK KGPAPKMLGN ELCSVCGDKA SGFHYNVLSC EGCKGFFRRS VIKGAHYICH SGGHCPMDTY MRRKCQECRL RKCRQAGMRE ECVLSEEQIR LKKLKRQEEE QAHATSLPPR ASSPPQILPQ LSPEQLGMIE KLVAAQQQCN RRSFSDRLRV TPWPMAPDPH SREARQQRFA HFTELAIVSV QEIVDFAKQL PGFLQLSRED QIALLKTSAI EVMLLETSRR YNPGSESITF LKDFSYNRED FAKAGLQVEF INPIFEFSRA MNELQLNDAE FALLIAISIF SADRPNVQDQ LQVERLQHTY VEALHAYVSI HHPHDRLMFP RMLMKLVSLR TLSSVHSEQV FALRLQDKKL PPLLSEIWDV HE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.