missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Paraneoplastic antigen-like protein 1

Product Code. 16022001
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16022001

Brand: enQuireBio™ QP7986ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Paraneoplastic antigen-like protein 5, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001096620.1
Gene ID (Entrez) 114824
Name Paraneoplastic antigen-like protein 1
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol PNMA5
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence TLLEDWCKGM DMDPRKALLI VGIPMECSEV EIQDTVKAGL QPLCAYRVLG RMFRREDNAK AVFIELADTV NYTTLPSHIP GKGGSWEVVV KPRNPDDEFL SRLNYFLKDE GRSMTDVARA LGCCSLPAES LDAEVMPQVR SPPLEPPKES MWYRKLKVFS GTASPSPGEE TFEDWLEQVT EIMPIWQVSE VEKRRRLLES LRGPALSIMR VLQANNDSIT VEQCLDALKQ IFGDKEDFRA SQFRFLQTSP KIGEKVSTFL LRLEPLLQKA VHKSPLSVRS TDMIRLKHLL ARVAMTPALR GKLELLDQRG CPPNFLELMK LIRDEEEWEN TEAVMKNKEK PSGRGRGASG RQARAEASVS APQATVQARS FSDSSPQTIQ GGLPPLVKRR RLLGSESTRG EDHGQATYPK AENQTPGREG PQAAGEELGN EAGAGAMSH
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.