missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PCMT1 Protein

A cDNA sequence encoding the PCMT1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12976-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942129

  • € 120.00 / 5µg

Estimated Shipment: 30-01-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PCMT1 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PCMT1 Human solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl, and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
RGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAWK SGGASHSELI HNLRKNGIIK TDKVFEVMLA TDRSHYAKCN PYMDSPQSIG FQATISAPHM HAYALELLFD QLHEGAKALD VGSGSGILTA CFARMVGCTG KVIGIDHIKE LVDDSINNVR KDDPTLLSSG RVQLVVGDGR MGYAEEAPYD AIHVGAAAPV VPQALIDQLK PGGRLILPVG PAGGNQMLEQ YDKLQDGSIK MKPLMGVIYV PLTDKEKQWS RWK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers