missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human PDGF-BB, Animal-Free Protein

Product Code. 18785396
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18785396

Brand: R&D Systems™ AFL220010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells

  • The ED50 for this effect is 1.5 to 6 ng/mL.
  • The specific activity of Recombinant Human PDGF-BB is approximately 5.8 x 103 IU/μg, which is calibrated against Recombinant Human PDGF-BB WHO International Standard a(NIBSC code: 94/728).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01127
For Use With (Application) Bioactivity
Gene ID (Entrez) 5155
Molecular Weight (g/mol) 12.3 kDa (monomer)
Quantity 10 μg
Source E. coli-derived human PDGF-BB protein Ser82-Thr190 Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol PDGFB
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 0.2 mg/mL in sterile 4 mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.