missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Recombinant Human PDGF-BB (CHO-expressed) Protein, CF

Product Code. 30058519
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
1mg
50µg
Product Code. Quantity unitSize
30058519 100 μg 100µg
30058545 10 μg 10µg
30058559 1 mg 1mg
30058549 50 μg 50µg
4 options
This item is not returnable. View return policy

Product Code. 30058519

Brand: R&DSystems™ 11565PB100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 2.50-25.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01127.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 5155
Molecular Weight (g/mol) M.W.-observed: 11-13 kDa, M.W.-theroretical: 12 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived human PDGF-BB protein Ser82-Thr190
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias c-sis;IBGC5;PDGF2;PDGF-2;PDGFB;Platelet-derived growth factor subunit B;SIS;SSV, PDGFBB
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in 4mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PDGF-BB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.