Learn More
enQuireBio™ Recombinant Human PDGFA Protein
Product Code. 15985839
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
Product Code. 15985839
Brand: enQuireBio™ QP528310ug
Recombinant Protein
Specifications
Accession Number NP_002598.4
Gene ID (Entrez) 5154
Name PDGFA Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol PDGFA
Biological Activity Shows bioactivity at a concentration of 50 ng/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 2.0 x 10^4 units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.