missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDZD11 / PDZK11 / PISP Protein
A DNA sequence encoding the Homo sapiens (Human) PDZD11 / PDZK11 / PISP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7680-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057568.1 | |
PDZD11 / PDZK11 / PISP Protein | |
Research Use Only | |
PDZD11 | |
Recombinant Protein | |
E. coli | |
MDSRIPYDDY PVVFLPAYEN PPAWIPPHER VHHPDYNNEL TQFLPRTITL KKPPGAQLGF NIRGGKASQL GIFISKVIPD SDAHRAGLQE GDQVLAVNDV DFQDIEHSKA VEILKTAREI SMRVRFFPYN YHRQKERTVH | |
Greater than 90% as determined by SDS-PAGE. |
51248 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |