missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Peroxiredoxin 2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-198 of Mouse Peroxiredoxin 2alpha The Recombinant Mouse Peroxiredoxin 2 Protein is derived from E. coli. The Recombinant Mouse Peroxiredoxin 2 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
| Concentration | 1 mg/ml |
| For Use With (Application) | Bioactivity, Functional Study, SDS-PAGE |
| Formulation | PBS (pH 7.4) containing 10% glycerol. |
| Gene ID (Entrez) | 7001 |
| Molecular Weight (g/mol) | 24.3 kDa |
| Quantity | 100 μg |
| Source | E. coli |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Gene Symbol | PRDX2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction