missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PHLDA2 Protein
A DNA sequence encoding the Homo sapiens (Human) PHLDA2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7664-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003302.1 | |
PHLDA2 Protein | |
Research Use Only | |
PHLDA2 | |
Recombinant Protein | |
E. coli | |
MKSPDEVLRE GELEKRSDSL FQLWKKKRGV LTSDRLSLFP ASPRARPKEL RFHSILKVDC VERTGKYVYF TIVTTDHKEI DFRCAGESCW NAAIALALID FQNRRALQDF RSRQERTAPA APAEDAVAAA AAAPSEPSEP SRPSPQPKPR TP | |
Greater than 80% as determined by SDS-PAGE. |
7262 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |