missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phospholipase D4 Protein
A DNA sequence encoding the Homo sapiens (Human) Phospholipase D4, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_620145.2 |
---|---|
Gene ID (Entrez) | 122618 |
Name | Phospholipase D4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011451
|
enQuireBio™
QP7894-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031451
|
enQuireBio™
QP7894-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021451
|
enQuireBio™
QP7894-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041451
|
enQuireBio™
QP7894-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051451
|
enQuireBio™
QP7894-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061451
|
enQuireBio™
QP7894-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_620145.2 | |
Phospholipase D4 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
122618 | |
Research Use Only | |
PLD4 | |
Recombinant Protein | |
E. coli | |
WQVPRPPTWG QVQPKDVPRS WEHGSSPAWE PLEAEARQQR DSCQLVLVES IPQDLPSAAG SPSAQPLGQA WLQLLDTAQE SVHVASYYWS LTGPDIGVND SSSQLGEALL QKLQQLLGRN ISLAVATSSP TLARTSTDLQ VLAARGAHVR QVPMGRLTRG VLHSKFWVVD GRHIYMGSAN MDWRSLTQVK ELGAVIYNCS HLAQDLEKTF QTYWVLGVPK AVLPKTWPQN FSSHFNRFQP FHGLFDGVPT TAYFSASPPA LCPQGRTRDL EALLAVMGSA QEFIYASVME YFPTTRFSHP PRYWPVLDNA LRAAAFGKGV RVRLLVGCGL NTDPTMFPYL RSLQALSNPA ANVSVDVKVF IVPVGNHSNI PFSRVNHSKF MVTEKAAYIG TSNWSEDYFS STAGVGLVVT QSPGAQPAGA TVQEQLRQLF ERDWSSRYAV GLDGQAPGQD CVWQG | |
Greater than 90% as determined by SDS-PAGE. |