missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PKC-a Protein

Product Code. 15922399
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
This item is not returnable. View return policy

Product Code. 15922399

Brand: enQuireBio™ QP130642ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PKC-a was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 5578
Name PKC-a Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol PRKCA
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence MADVYPGNDS TASQDVANRF ARKGALRQKN VHEVKDHKFI ARFFKQPTFC SHCTDFIWGF GKQGFQCQVC CFVVHKRCHE FVTFSCPGAD KGPDTDDPRS KHKFKIHTYG SPTFCDHCGS LLYGLIHQGM KCDTCDMNVH KQCVINVPSL CGMDHTEKRG RIYLKAEVAD EKLHVTVRDA KNLIPMDPNG LSDPYVKLKL IPDPKNESKQ KTKTIRSTLN PQWNESFTFK LKPSDKDRRL SVEIWDWDRT TRNDFMGSLS FGVSELMKMP ASGWYKLLNQ EEGEYYNVPI PEGDEEGNME LRQKFEKAKL GPAGNKVISP SEDRKQPSNN LDRVKLTDFN FLMVLGKGSF GKVMLADRKG TEELYAIKIL KKDVVIQDDD VECTMVEKRV LALLDKPPFL TQLHSCFQTV DRLYFVMEYV NGGDLMYHIQ QVGKFKEPQA VFYAAEISIG LFFLHKRGII YRDLKLDNVM LDSEGHIKIA DFGMCKEHMM DGVTTRTFCG TPDYIAPEII AYQPYGKSVD WWAYGVLLYE MLAGQPPFDG EDEDELFQSI MEHNVSYPKS LSKEAVSICK GLMTKHPAKR LGCGPEGERD VREHAFFRGI DWEKLENREI QPPFKPKVCG KGAENFDKFF TRGQPVLTPP DQLVIANIDQ SDFEGFSYVN PQFVHPILQS AV
Buffer PKC-a is supplied in 20mM HEPES pH 7.5, 250mM NaCl, 50% glycerol, 2mM EDTA, 2mM EGTA, 0.05%Triton X-100 and 5mM DTT.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.