missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PNRC2 Protein
A cDNA sequence encoding the PNRC2 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 4090.00
Specifications
Name | PNRC2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942469
|
enQuireBio™
QP13087-5UG |
5 μg |
€ 137.00
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932469
|
enQuireBio™
QP13087-20UG |
20 μg |
€ 212.00
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922469
|
enQuireBio™
QP13087-1MG |
1 mg |
€ 4090.00
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PNRC2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PNRC2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGGGERY NIPAPQSRNV SKNQQQLNRQ KTKEQNSQMK IVHKKKERGH GYNSSAAAWQ AMQNGGKNKN FPNNQSWNSS LSGPRLLFKS QANQNYAGAK FSEPPSPSVL PKPPSHWVPV SFNPSDKEIM TFQLKTLLKV QV | |
Greater than 90% as determined by SDS-PAGE. |