Learn More
enQuireBio™ Recombinant Human POLR2J Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | POLR2J Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMNAPPAF ESFLLFEGEK KITINKDTKV PNACLFTINK EDHTLGNIIK SQLLKDPQVL FAGYKVPHPL EHKIIIRVQT TPDYSPQEAF TNAITDLISE LSLLEERFRV AIKDKQEGIE |
| Buffer | POLR2J protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol, 5mM DTT and 2mM EDTA. |
| Show More |