missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PREP Protein

Product Code. 15992609
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
This item is not returnable. View return policy

Product Code. 15992609

Brand: enQuireBio™ QP131362ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PREP was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 5550
Name PREP Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol PREP
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMLSLQYP DVYRDETAVQ DYHGHKICDP YAWLEDPDSE QTKAFVEAQN KITVPFLEQC PIRGLYKERM TELYDYPKYS CHFKKGKRYF YFYNTGLQNQ RVLYVQDSLE GEARVFLDPN ILSDDGTVAL RGYAFSEDGE YFAYGLSASG SDWVTIKFMK VDGAKELPDV LERVKFSCMA WTHDGKGMFY NSYPQQDGKS DGTETSTNLH QKLYYHVLGT DQSEDILCAE FPDEPKWMGG AELSDDGRYV LLSIREGCDP VNRLWYCDLQ QESSGIAGIL KWVKLIDNFE GEYDYVTNEG TVFTFKTNRQ SPNYRVINID FRDPEESKWK VLVPEHEKDV LEWIACVRSN FLVLCYLHDV KNILQLHDLT TGALLKTFPL DVGSIVGYSG QKKDTEIFYQ FTSFLSPGII YHCDLTKEEL EPRVFREVTV KGIDASDYQT VQIFYPSKDG TKIPMFIVHK KGIKLDGSHP AFLYGYGGFN ISITPNYSVS RLIFVRHMGG ILAVANIRGG GEYGETWHKG GILANKQNCF DDFQCAAEYL IKEGYTSPKR LTINGGSNGG LLVAACANQR PDLFGCVIAQ VGVMDMLKFH KYTIGHAWTT DYGCSDSKQH FEWLVKYSPL HNVKLPEADD IQYPSMLLLT ADHDDRVVPL HSLKFIATLQ YIVGRSRKQS NPLLIHVDTK AGHGAGKPTA KVIEEVSDMF AFIARCLNVD WIP
Buffer PREP protein solution (0.25 mg/ml) containing PBS buffer (pH 7.4), 30% glycerol and 1mM DTT.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.