missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRKAB2 Protein

A DNA sequence encoding the Homo sapiens (Human) PRKAB2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7495-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029000

Quantity Price
1 € 1180.00 / 500µg
3 € 1100.47 / 500µg
Estimated Shipment
03-11-2020
Add to basket


Specifications

Specifications

NP_005390.1
PRKAB2 Protein
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
5565
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PRKAB2
Recombinant Protein
E. coli
MGNTTSDRVS GERHGAKAAR SEGAGGHAPG KEHKIMVGST DDPSVFSLPD SKLPGDKEFV SWQQDLEDSV KPTQQARPTV IRWSEGGKEV FISGSFNNWS TKIPLIKSHN DFVAILDLPE GEHQYKFFVD GQWVHDPSEP VVTSQLGTIN NLIHVKKSDF EVFDALKLDS MESSETSCRD LSSSPPGPYG QEMYAFRSEE RFKSPPILPP HLLQVILNKD TNISCDPALL PEPNHVMLNH LYALSIKDSV MVLSATHRYK KKYVTTLLYK PI
Videos
SDS
Documents