missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Probable G-protein coupled receptor 75 Protein

A DNA sequence encoding the Homo sapiens (Human) Probable G-protein coupled receptor 75, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6113-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030890

  • € 2385.00 / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006785.1
Probable G-protein coupled receptor 75 Protein
Research Use Only
GPR75
Recombinant Protein
E. coli
NPFIYSRNSA GLRRKVLWCL QYIGLGFFCC KQKTRLRAMG KGNLEVNRNK SSHHETNSAY MLSPKPQKKF VDQACGPSHS KESMVSPKIS AGHQHCGQSS STPINTRIEP YYSIYNSSPS QEESSPCNLQ PVNSFGFANS YIAMHYHTTN DLVQEYDSTS AKQIPVPSV
Greater than 90% as determined by SDS-PAGE.
10936
1 mg
Not determined.
Cytoplasmic Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers