missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proteasomal ubiquitin receptor ADRM1 Protein

A DNA sequence encoding the Homo sapiens (Human) Proteasomal ubiquitin receptor ADRM1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7545-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029300

Quantity Price
1 € 1715.00 / 500µg
3 € 1600.44 / 500µg
Estimated Shipment
23-10-2020
Add to basket


Specifications

Specifications

NP_001268366.1
Proteasomal ubiquitin receptor ADRM1 Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
11047
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADRM1
Recombinant Protein
E. coli
TTSGALFPSL VPGSRGASNK YLVEFRAGKM SLKGTTVTPD KRKGLVYIQQ TDDSLIHFCW KDRTSGNVED DLIIFPDDCE FKRVPQCPSG RVYVLKFKAG SKRLFFWMQE PKTDQDEEHC RKVNEYLNNP PMPGALGASG SSGHELSALG GEGGLQSLLG NMSHSQLMQL IGPAGLGGLG GLGALTGPGL ASLLGSSGPP GSSSSSSSRS QSAAVTPSST TSSTRATPAP SAPAAASATS PSPAPSSGNG ASTAASPTQP IQLSDLQSIL ATMNVPAGPA GGQQVDLASV LTPEIMAPIL ANADVQERLL PYLPSGESLP QTADEIQNTL TSPQFQQALG MFSAALASGQ LGPLMCQFGL PAEAVEAANK GDVEAFAKAM QNNAKPEQKE GDTKDKKDEE EDMSLD
Videos
Documents