missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Proteasome 20S beta2 His Protein
Shop All Bio Techne Products
Click to view available options
:
50μg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-201 of Human Proteasome 20S beta2 The Recombinant Human Proteasome 20S beta2 Protein is derived from E. coli. The Recombinant Human Proteasome 20S beta2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | P49721 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 30% glycerol, 0.1M NaCl |
| Gene ID (Entrez) | 5690 |
| Molecular Weight (g/mol) | 24.9kDa |
| Name | Proteasome 20S beta2 Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction