missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein FAM46C Protein
A DNA sequence encoding the Homo sapiens (Human) Protein FAM46C, was expressed in the hosts and tags indicated.
€ 287.00 - € 3175.00
Specifications
Accession Number | NP_060179.2 |
---|---|
Gene ID (Entrez) | 54855 |
Name | Protein FAM46C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16080481
|
enQuireBio™
QP7730-YE-100UG |
100 μg |
€ 920.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020491
|
enQuireBio™
QP7730-YE-500UG |
500 μg |
€ 2650.00
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090481
|
enQuireBio™
QP7730-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010491
|
enQuireBio™
QP7730-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030491
|
enQuireBio™
QP7730-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000491
|
enQuireBio™
QP7730-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_060179.2 | |
Protein FAM46C Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
54855 | |
Research Use Only | |
FAM46C | |
Recombinant Protein | |
Yeast | |
MAEESSCTRD CMSFSVLNWD QVSRLHEVLT EVVPIHGRGN FPTLEITLKD IVQTVRSRLE EAGIKVHDVR LNGSAAGHVL VKDNGLGCKD LDLIFHVALP TEAEFQLVRD VVLCSLLNFL PEGVNKLKIS PVTLKEAYVQ KLVKVCTDTD RWSLISLSNK NGKNVELKFV DSIRRQFEFS VDSFQIILDS LLFFYDCSNN PISEHFHPTV IGESMYGDFE EAFDHLQNRL IATKNPEEIR GGGLLKYSNL LVRDFRPTDQ EEIKTLERYM CSRFFIDFPD ILEQQRKLET YLQNHFAEEE RSKYDYLMIL RRVVNESTVC LMGHERRQTL NLISLLALRV LAEQNIIPSA TNVTCYYQPA PYVSDGNFSN YYVAHPPVTY SQPYPTWLPC N | |
Greater than 90% as determined by SDS-PAGE. |