missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein FAM46C Protein

A DNA sequence encoding the Homo sapiens (Human) Protein FAM46C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7730-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030491

Quantity Price
1 € 416.00 / 50µg
6 € 388.50 / 50µg
Estimated Shipment
03-11-2020
Add to basket


Specifications

Specifications

NP_060179.2
Protein FAM46C Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
54855
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FAM46C
Recombinant Protein
Yeast
MAEESSCTRD CMSFSVLNWD QVSRLHEVLT EVVPIHGRGN FPTLEITLKD IVQTVRSRLE EAGIKVHDVR LNGSAAGHVL VKDNGLGCKD LDLIFHVALP TEAEFQLVRD VVLCSLLNFL PEGVNKLKIS PVTLKEAYVQ KLVKVCTDTD RWSLISLSNK NGKNVELKFV DSIRRQFEFS VDSFQIILDS LLFFYDCSNN PISEHFHPTV IGESMYGDFE EAFDHLQNRL IATKNPEEIR GGGLLKYSNL LVRDFRPTDQ EEIKTLERYM CSRFFIDFPD ILEQQRKLET YLQNHFAEEE RSKYDYLMIL RRVVNESTVC LMGHERRQTL NLISLLALRV LAEQNIIPSA TNVTCYYQPA PYVSDGNFSN YYVAHPPVTY SQPYPTWLPC N
Videos
SDS
Documents