missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein Hikeshi Protein
A DNA sequence encoding the Homo sapiens (Human) Protein Hikeshi, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_057485.2 |
---|---|
Gene ID (Entrez) | 51501 |
Name | Protein Hikeshi Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16090121
|
enQuireBio™
QP7678-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010131
|
enQuireBio™
QP7678-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000131
|
enQuireBio™
QP7678-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020131
|
enQuireBio™
QP7678-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030131
|
enQuireBio™
QP7678-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040131
|
enQuireBio™
QP7678-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_057485.2 | |
Protein Hikeshi Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
51501 | |
Research Use Only | |
C11orf73 | |
Recombinant Protein | |
E. coli | |
MFGCLVAGRL VQTAAQQVAE DKFVFDLPDY ESINHVVVFM LGTIPFPEGM GGSVYFSYPD SNGMPVWQLL GFVTNGKPSA IFKISGLKSG EGSQHPFGAM NIVRTPSVAQ IGISVELLDS MAQQTPVGNA AVSSVDSFTQ FTQKMLDNFY NFASSFAVSQ AQMTPSPSEM FIPANVVLKW YENFQRRLAQ NPLFWKT | |
Greater than 90% as determined by SDS-PAGE. |