missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein Hikeshi Protein
A DNA sequence encoding the Homo sapiens (Human) Protein Hikeshi, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7678-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057485.2 | |
Protein Hikeshi Protein | |
Research Use Only | |
C11orf73 | |
Recombinant Protein | |
E. coli | |
MFGCLVAGRL VQTAAQQVAE DKFVFDLPDY ESINHVVVFM LGTIPFPEGM GGSVYFSYPD SNGMPVWQLL GFVTNGKPSA IFKISGLKSG EGSQHPFGAM NIVRTPSVAQ IGISVELLDS MAQQTPVGNA AVSSVDSFTQ FTQKMLDNFY NFASSFAVSQ AQMTPSPSEM FIPANVVLKW YENFQRRLAQ NPLFWKT | |
Greater than 90% as determined by SDS-PAGE. |
51501 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |