missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein SSX2 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein SSX2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7535-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001157889.1 | |
Protein SSX2 Protein | |
Research Use Only | |
SSX2 | |
Recombinant Protein | |
E. coli | |
MNGDDAFARR PTVGAQIPEK IQKAFDDIAK YFSKEEWEKM KASEKIFYVY MKRKYEAMTK LGFKATLPPF MCNKRAEDFQ GNDLDNDPNR GNQVERPQMT FGRLQGISPK IMPKKPAEEG NDSEEVPEAS GPQNDGKELC PPGKPTTSEK IHERSGPKRG EHAWTHRLRE RKQLVIYEEI SDPEEDDE | |
Greater than 90% as determined by SDS-PAGE. |
6757 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |