missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&DSystems™ Recombinant Human PSMA/FOLH1/NAALADase I Avi Fc Protein, CF
Click to view available options
:
20 μg
Description
Measured by its ability to hydrolyze the substrate N-acetyl-L-Asp-L-Glu into N-acetyl-L-Asp and L-Glu. The L-Glu product is measured by fluorescence after its derivatization by ortho-phthaldialdehyde. The specific activity is >325 pmol/min/μg, as measured under the described conditions. Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human PSMA/FOLH1/NAALADase I Avi-tag Fc Chimera (Catalog # AVI11535) binds Human PSMA/FOLH1/NAALADase I Antibody (Catalog # MAB4234 ) with an ED50 of 0.0750-1.20 μg/mL.
Specifications
Specifications
| Accession Number | Q04609.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 2346 |
| Molecular Weight (g/mol) | M.W.-observed: 112-125 kDa, under reducing conditions., M.W.-theroretical: 108 kDa |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human PSMA/FOLH1/NAALADase I protein Avi-tag MD Human IgG1 (Pro100-Lys330) IEGR Human NAALADase-1 (Lys44-Ala750) (N-terminus) Accession # Q04609.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, FGCP, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLH1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPII, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, mopsm, NAALAD1, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, NAALADase I, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction