missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTMS Protein

A cDNA sequence encoding the PTMS was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13207-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932839

  • € 137.00 / 2µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

5763
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSEKSVE AAAELSAKDL KEKKEKVEEK ASRKERKKEV VEEEENGAEE EEEETAEDGE EEDEGEEEDE EEEEEDDEGP ALKRAAEEED EADPKRQKTE NGASA
Greater than 95.0% as determined by SDS-PAGE.
PTMS Protein
Research Use Only
PTMS
Human
His
PTMS protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers