missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTPN1 Protein

A cDNA sequence encoding the PTPN1 was constructed and used to recombinantly synthesize the protein.

€ 137.00 - € 2725.00

Specifications

Name PTPN1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Specific activity is > 10,000 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmole of p-nitrophenyl phosphate (pNPP) per minute at pH 7.5 at 37C.
Product Type Recombinant Protein
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15925378
View Documents Promotion Details
enQuireBio™
QP10856-10UG
10 μg
€ 137.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15945378
View Documents Promotion Details
enQuireBio™
QP10856-50UG
50 μg
€ 212.00
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15935378
View Documents Promotion Details
enQuireBio™
QP10856-1MG
1 mg
€ 2725.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

PTPN1 Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MEMEKEFEQI DKSGSWAAIY QDIRHEASDF PCRVAKLPKN KNRNRYRDVS PFDHSRIKLHQEDNDYINAS LIKMEEAQRS YILTQGPLPN TCGHFWEMVW EQKSRGVVML NRVMEKGSLKCAQYWPQKEE KEMIFEDTNL KLTLISEDIK SYYTVRQLEL ENLTTQETRE ILHFHYTTWPDFGVPESPAS FLNFLFKVRE SGSLSPEHGP VVVHCSAGIG RSGTFCLADT CLLLMDKRKDPSSVDIKKVL LEMRKFRMGL IQTADQLRFS YLAVIEGAKF IMGDSSVQDQ WKELSHEDLE PPPEHIPPPPRPPKRILEPHN
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Specific activity is > 10,000 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmole of p-nitrophenyl phosphate (pNPP) per minute at pH 7.5 at 37C.
Human
Untagged
The protein (1 mg/ml) contains 25mM Tris-HCl, pH 7.5, 2mM beta-mercaptoethanol, 1mM EDTA, 1mM DTT and 20% Glycerol.
Videos
SDS
Documents

Documents

Product Certifications