missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PTPRD Fc Chimera Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human PTPRD Fc Chimer is immobilized at 1 μg/mL, 100 μL/well, Recombinant Human IL-1 RAPL1/IL-1 R8 Fc Chimera (Catalog # 9949-ML ) binds with an ED50 of 1.5-9 ng/mL.
Specifications
Specifications
| Accession Number | P23468 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 5789 |
| Molecular Weight (g/mol) | M.W. (Observed): 137-151 kDa, reducing conditions; M.W. (theoretical): 154 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human PTPRD protein Human PTPRD (Glu21-Ser1174) (N-terminus) Accession # P23468 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?