missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTPRN Protein

A DNA sequence encoding the Homo sapiens (Human) PTPRN, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9748-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16078641

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001186692.1
PTPRN Protein
Research Use Only
PTPRN
Recombinant Protein
Yeast
VSAHGCLFDR RLCSHLEVCI QDGLFGQCQV GVGQARPLLQ VTSPVLQRLQ GVLRQLMSQG LSWHDDLTQY VISQEMERIP RLRPPEPRPR DRSGLAPKRP GPAGELLLQD IPTGSAPAAQ HRLPQPPVGK GGAGASSSLS PLQAELLPPL LEHLLLPPQP PHPSLSYEPA LLQPYLFHQF GSRDGSRVSE GSPGMVSVGP LPKAEAPALF SRTASKGIFG DHPGHSYGDL PGPSPAQLFQ DSGLLYLAQE LPAPSRARVP RLPEQGSSSR AEDSPEGYEK EGLGDRGEKP ASPAVQPDAA LQRLAAVLAG YGVELRQLTP EQLSTLLTLL QLLPKGAGRN PGGVVNVGAD IKKTMEGPVE GRDTAELPAR TSPMPGHPTA SPTSSEVQQV PSPVSSEPPK AARPPVTPVL LEKKSPLGQS QPTVAGQPSA RPAAEEYGYI VTDQKPLSLA AGVKLLEILA EHVHMSSGSF INISVVGPAL TFRIRHNEQN LSLADVTQQA GLVKSELEAQ TGLQILQTGV GQREEAAAVL PQTAHSTSPM R
Greater than 90% as determined by SDS-PAGE.
5798
50 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers