missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ptprn2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ptprn2, was expressed in the hosts and tags indicated.
€ 202.00 - € 2765.00
Specifications
Accession Number | NP_001295196.1 |
---|---|
Gene ID (Entrez) | 5799 |
Name | Ptprn2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088881
|
enQuireBio™
QP9873-YE-100UG |
100 μg |
€ 645.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008891
|
enQuireBio™
QP9873-YE-1MG |
1 mg |
€ 2765.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098881
|
enQuireBio™
QP9873-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018891
|
enQuireBio™
QP9873-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028891
|
enQuireBio™
QP9873-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038891
|
enQuireBio™
QP9873-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001295196.1 | |
Ptprn2 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5799 | |
Research Use Only | |
Ptprn2 | |
Recombinant Protein | |
Yeast | |
AAPSSVPRGR QLPGRLGCLL EEGLCGASEA CVNDGVFGRC QKVPAMDFYR YEVSPVALQR LRVALQKLSG TGFTWQDDYT QYVMDQELAD LPKTYLRRPE ASSPARPSKH SVGSERRYSR EGGAALANAL RRHLPFLEAL SQAPASDVLA RTHTAQDRPP AEGDDRFSES ILTYVAHTSA LTYPPGSRTQ LREDLLPRTL GQLQPDELSP KVDSGVDRHH LMAALSAYAA QRPPAPPGEG SLEPQYLLRA PSRMPRPLLA PAAPQKWPSP LGDSEDPSST GDGARIHTLL KDLQRQPAEV RGLSGLELDG MAELMAGLMQ GVDHGVARGS PGRAALGESG EQADGPKATL RGDSFPDDGV QDDDDRLYQE VHRLSATLGG LLQDHGSRLL PGALPFARPL DMERKKSEHP ESSLSSEEET AGVENVKSQT YSKDLLGQQP HSEPGAAAFG ELQNQMPGPS KEEQSLPAGA QEALSDGLQL EVQPSEEEAR GYIVTDRDPL RPEEGRRLVE DVARLLQVPS SAFADVEVLG PAVTFKVSAN VQNVTTEDVE KATVDNKDKL EETSGLKILQ TGVGSKSKLK FLPPQAEQED STKF | |
Greater than 90% as determined by SDS-PAGE. |