missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human QPRT Protein
A cDNA sequence encoding the QPRT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13225-1mg
Additional Details : Weight : 0.01000kg
Specifications
QPRT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDAEGLALLL PPVTLAALVD SWLREDCPGL NYAALVSGAG PSQAALWAKS PGVLAGQPFF DAIFTQLNCQ VSWFLPEGSK LVPVARVAEV RGPAHCLLLG ERVALNTLAR CSGIASAAAA AVEAARGAGW TGHVAGTRKT TPGFRLVEKY GLLVGGAASH RYDLGGLVMV KDNHVVAAGG VEKAVRAARQ AADFALKVEV ECSSLQEAVQ AAEAGADLVL LDNFKPEELH PTATVLKAQF PSVAVEASGG ITLDNLPQFC GPHIDVISMG MLTQAAPALD FSLKLFAKEV APVPKIH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The QPRT solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |