missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human QSOX1/Quiescin Q6 Protein
Click to view available options
:
Recombinant Human QSOX1/Quiescin Q6 Protein, Carrier-Free
Description
The Recombinant Human QSOX1/Quiescin Q6 Protein is derived from E. coli. The Recombinant Human QSOX1/Quiescin Q6 Protein has been validated for the following applications: Enzyme Activity.
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| For Use With (Application) | Enzyme Activity |
| Source | E. coli-derived human QSOX1/Quiescin Q6 protein Ser33-Ala546,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction