Learn More
enQuireBio™ Recombinant Human RAET1E Protein
A cDNA sequence encoding the RAET1E was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 7875.00
Specifications
Name RAET1E Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15932989
View Documents Promotion Details
enQuireBio™
QP13254-HIS-1UG
1 μg
€ 137.00
1µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
15942989
View Documents Promotion Details
enQuireBio™
QP13254-HIS-2UG
2 μg
€ 137.00
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
15962989
View Documents Promotion Details
enQuireBio™
QP13254-HIS-5UG
5 μg
€ 212.00
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
15912989
View Documents Promotion Details
enQuireBio™
QP13254-HIS-10UG
10 μg
€ 212.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
15952989
View Documents Promotion Details
enQuireBio™
QP13254-HIS-50UG
50 μg
€ 1815.00
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
15922989
View Documents Promotion Details
enQuireBio™
QP13254-HIS-1MG
1 mg
€ 7875.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Specifications
RAET1E Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RAET1E protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Research Use Only
Recombinant Protein
Baculovirus
ADPHSLCFNF TIKSLSRPGQ PWCEAQVFLN KNLFLQYNSD NNMVKPLGLL GKKVYATSTW GELTQTLGEV GRDLRMLLCD IKPQIKTSDP STLQVEMFCQ REAERCTGAS WQFATNGEKS LLFDAMNMTW TVINHEASKI KETWKKDRGL EKYFRKLSKG DCDHWLREFL GHWEAMPEPT VSPVNASDIH WSSSSLPDVE PKSCDKTHTC PPCPAPELLG GPSVFLFPPK PKDTLMISRT PEVTCVVVDV SHEDPEVKFN WYVDGVEVHN AKTKPREEQY NSTYRVVSVL TVLHQDWLNG KEYKCKVSNK ALPAPIEKTI SKAKGQPREP QVYTLPPSRD ELTKNQVSLT CLVKGFYPSD IAVEWESNGQ PENNYKTTPP VLDSDGSFFL YSKLTVDKSR WQQGNVFSCS VMHEALHNHY TQKSLSLSPG KHHHHHH
Greater than 85.0% as determined by SDS-PAGE.