missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ragulator complex protein LAMTOR1 Protein
A DNA sequence encoding the Homo sapiens (Human) Ragulator complex protein LAMTOR1, was expressed in the hosts and tags indicated.
€ 245.00 - € 3405.00
Specifications
Accession Number | NP_060377.1 |
---|---|
Gene ID (Entrez) | 55004 |
Name | Ragulator complex protein LAMTOR1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16010671
|
enQuireBio™
QP7766-EC-100UG |
100 μg |
€ 765.00
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030671
|
enQuireBio™
QP7766-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020671
|
enQuireBio™
QP7766-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040671
|
enQuireBio™
QP7766-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050671
|
enQuireBio™
QP7766-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060671
|
enQuireBio™
QP7766-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_060377.1 | |
Ragulator complex protein LAMTOR1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55004 | |
Research Use Only | |
LAMTOR1 | |
Recombinant Protein | |
E. coli | |
MGCCYSSENE DSDQDREERK LLLDPSSPPT KALNGAEPNY HSLPSARTDE QALLSSILAK TASNIIDVSA ADSQGMEQHE YMDRARQYST RLAVLSSSLT HWKKLPPLPS LTSQPHQVLA SEPIPFSDLQ QVSRIAAYAY SALSQIRVDA KEELVVQFGI P | |
Greater than 90% as determined by SDS-PAGE. |