missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras association domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Ras association domain-containing protein 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7834-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_872604.1 | |
Ras association domain-containing protein 1 | |
Research Use Only | |
RASSF5 | |
Recombinant Protein | |
E. coli | |
MTVDSSMSSG YCSLDEELED CFFTAKTTFF RNAQSKHLSK NVCKPVEETQ RPPTLQEIKQ KIDSYNTREK NCLGMKLSED GTYTGFIKVH LKLRRPVTVP AGIRPQSIYD AIKEVNLAAT TDKRTSFYLP LDAIKQLHIS STTTVSEVIQ GLLKKFMVVD NPQKFALFKR IHKDGQVLFQ KLSIADRPLY LRLLAGPDTE VLSFVLKENE TGEVEWDAFS IPELQNFLTI LEKEEQDKIQ QVQKKYDKFR QKLEEALRES QGKPG | |
Greater than 80% as determined by SDS-PAGE. |
83593 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |