missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RBBP9 Protein

A cDNA sequence encoding the RBBP9 was constructed and used to recombinantly synthesize the protein.

€ 137.00 - € 4090.00

Specifications

Name RBBP9 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15953059
View Documents Promotion Details
enQuireBio™
QP13277-5UG
5 μg
€ 137.00
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15943059
View Documents Promotion Details
enQuireBio™
QP13277-20UG
20 μg
€ 212.00
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15933059
View Documents Promotion Details
enQuireBio™
QP13277-1MG
1 mg
€ 4090.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

RBBP9 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RBBP9 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MASPSKAVIV PGNGGGDVTT HGWYGWVKKE LEKIPGFQCL AKNMPDPITA RESIWLPFME TELHCDEKTI IIGHSSGAIA AMRYAETHRV YAIVLVSAYT SDLGDENERA SGYFTRPWQW EKIKANCPYI VQFGSTDDPF LPWKEQQEVA DRLETKLHKF TDCGHFQNTE FHELITVVKS LLKVPA
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications