missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RCAN1 Protein

A cDNA sequence encoding the RCAN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13286-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943089

  • € 137.00 / 5µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RCAN1 Protein
Research Use Only
Recombinant Protein
E. coli
MKLYFAQTLH IGSSHLAPPN PDKQFLISPP ASPPVGWKQV EDATPVINYD LLYAISKLGPGEKYELHAAT DTTPSVVVHV CESDQEKEEE EEMERMRRPK PKIIQTRRPE YTPIHLS
Greater than 90.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein solution (1 mg/ml) contains 20mM Tris-HCl pH 7.5, 100mM NaCl and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers