missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human REN Protein

A cDNA sequence encoding the REN was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13298-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933129

  • € 181.00 / 5µg

Estimated Shipment: 02-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

REN Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
REN protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Greater than 90.0% as determined by analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
LPTDTTTFKR IFLKRMPSIR ESLKERGVDM ARLGPEWSQP MKRLTLGNTT SSVILTNYMD TQYYGEIGIG TPPQTFKVVF DTGSSNVWVP SSKCSRLYTA CVYHKLFDAS DSSSYKHNGT ELTLRYSTGT VSGFLSQDII TVGGITVTQM FGEVTEMPAL PFMLAEFDGV VGMGFIEQAI GRVTPIFDNI ISQGVLKEDV FSFYYNRDSE NSQSLGGQIV LGGSDPQHYE GNFHYINLIK TGVWQIQMKG VSVGSSTLLC EDGCLALVDT GASYISGSTS SIEKLMEALG AKKRLFDYVV KCNEGPTLPD ISFHLGGKEY TLTSADYVFQ ESYSSKKLCT LAIHAMDIPP PTGPTWALGA TFIRKFYTEF DRRNNRIGFA LARLEHHHHH H
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers