missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Reticulocalbin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Reticulocalbin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9756-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028671

  • € 491.00 / 100µg


Estimated Shipment
03-11-2020
Add to basket

Specifications

Specifications

NP_002892.1
Reticulocalbin-1 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
5954
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RCN1
Recombinant Protein
Yeast
PTVRKERVVR PDSELGERPP EDNQSFQYDH EAFLGKEDSK TFDQLTPDES KERLGKIVDR IDNDGDGFVT TEELKTWIKR VQKRYIFDNV AKVWKDYDRD KDDKISWEEY KQATYGYYLG NPAEFHDSSD HHTFKKMLPR DERRFKAADL NGDLTATREE FTAFLHPEEF EHMKEIVVLE TLEDIDKNGD GFVDQDEYIA DMFSHEENGP EPDWVLSERE QFNEFRDLNK DGKLDKDEIR HWILPQDYDH AQAEARHLVY ESDKNKDEKL TKEEILENWN MFVGSQATNY GEDLTKNHDE L
Videos
SDS
Documents