missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ Recombinant Human Ring finger protein 138 GST (N-Term) Protein
Shop alle Bio Techne produkter
Klik for at se tilgængelige muligheder
Quantity:
10 μg
25 μg
Pakningsstørrelse:
10µg
25µg
Beskrivelse
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Tekniske data
Tekniske data
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 51444 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 52.58 kDa |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | E3 ubiquitin-protein ligase RNF138, EC 6.3.2.-, hNARF, HSD-4, MGC8758, NARF, Nemo-like kinase-associated RING finger protein, ring finger protein 138NLK-associated RING finger protein, STRIN |
| Gene Symbol | RNF138 |
| Research Category | Zinc Finger |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion