missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human ROBO1/DUTT1 Fc Chimera Protein

Product Code. 18711258
Click to view available options
:
50μg
This item is not returnable. View return policy

Product Code. 18711258

Brand: R&D Systems™ 8975RB050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human ROBO1/DUTT1 Fc Chimera Protein is derived from NS0. The Recombinant Human ROBO1/DUTT1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 6091
Molecular Weight (g/mol) Theoretical M.W.: 119kDa
Quantity 50μg
Source Mouse myeloma cell line,NS0-derived human ROBO1 protein, (N-terminus) Human ROBO1/DUTT1 (Ser20-Pro858) Accession &Num; NP_598334.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias DUTT1, DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila)
Common Name ROBO1
Gene Symbol ROBO1
Biological Activity Measured by its binding ability in a functional ELISA. When recombinant human Slit3 is coated at 250ng/mL, Recombinant Human ROBO1 Fc Chimera binds with a typical ED50 of 100-600ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Human ROBO1/DUTT1 (Ser20-Pro858) Accession # NP_598334.1, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.