missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RRM2B / P53R2 Protein

A DNA sequence encoding the Homo sapiens (Human) RRM2B / P53R2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7745-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030551

Quantity Price
1 € 352.00 / 50µg
6 € 328.49 / 50µg
In stock, ready to ship: 103
Add to basket


Specifications

Specifications

NP_001165948.1
RRM2B / P53R2 Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
50484
Greater than 80% as determined by SDS-PAGE.
Research Use Only
RRM2B
Recombinant Protein
E. coli
MGDPERPEAA GLDQDERSSS DTNESEIKSN EEPLLRKSSR RFVIFPIQYP DIWKMYKQAQ ASFWTAEEVD LSKDLPHWNK LKADEKYFIS HILAFFAASD GIVNENLVER FSQEVQVPEA RCFYGFQILI ENVHSEMYSL LIDTYIRDPK KREFLFNAIE TMPYVKKKAD WALRWIADRK STFGERVVAF AAVEGVFFSG SFAAIFWLKK RGLMPGLTFS NELISRDEGL HCDFACLMFQ YLVNKPSEER VREIIVDAVK IEQEFLTEAL PVGLIGMNCI LMKQYIEFVA DRLLVELGFS KVFQAENPFD FMENISLEGK TNFFEKRVSE YQRFAVMAET TDNVFTLDAD F
Videos
Documents