missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A15 / S100A7A Protein

A DNA sequence encoding the Homo sapiens (Human) S100A15 / S100A7A, was expressed in the hosts and tags indicated.

€ 160.00 - € 2385.00

Specifications

Accession Number NP_789793.1
Gene ID (Entrez) 338324
Name S100A15 / S100A7A Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16090801
View Documents Promotion Details
enQuireBio™
QP7789-EC-100UG
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16010811
View Documents Promotion Details
enQuireBio™
QP7789-EC-1MG
1 mg
€ 2385.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16000811
View Documents Promotion Details
enQuireBio™
QP7789-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16020811
View Documents Promotion Details
enQuireBio™
QP7789-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16030811
View Documents Promotion Details
enQuireBio™
QP7789-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16040811
View Documents Promotion Details
enQuireBio™
QP7789-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_789793.1
S100A15 / S100A7A Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
338324
Research Use Only
S100A7A
Recombinant Protein
E. coli
SNTQAERSII GMIDMFHKYT GRDGKIEKPS LLTMMKENFP NFLSACDKKG IHYLATVFEK KDKNEDKKID FSEFLSLLGD IAADYHKQSH GAAPCSGGSQ
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications