Learn More
enQuireBio™ Recombinant Human Secretin Protein
Product Code. 15943599
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15943599 1 mg 1mg
15963599 5 mg 5mg
15953599 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15943599 Supplier enQuireBio™ Supplier No. QP134491mg
Recombinant Protein
Specifications
Name Secretin Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MAPRPLLLLL LLLGGSAARP APPRARRHSD GTFTSELSRL REGARLQRLL QGLVGKRSEQ DAENSMAWTR LSAGLLCPSG SNMPILQAWM PLDGTWSPWL PPGPMVSEPA GAAAEGTLRP R
Buffer The protein (1 mg/ml) was lyophilized with no additives.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.