missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Secretin Protein
Recombinant Protein
€ 125.00 - € 595.00
Specifications
Name | Secretin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943599
|
enQuireBio™
QP13449-1MG |
1 mg |
€ 125.00
1mg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963599
|
enQuireBio™
QP13449-5MG |
5 mg |
€ 212.00
5mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953599
|
enQuireBio™
QP13449-25MG |
25 mg |
€ 595.00
25mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Secretin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) was lyophilized with no additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAPRPLLLLL LLLGGSAARP APPRARRHSD GTFTSELSRL REGARLQRLL QGLVGKRSEQ DAENSMAWTR LSAGLLCPSG SNMPILQAWM PLDGTWSPWL PPGPMVSEPA GAAAEGTLRP R | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |