missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Semaphorin 3G Fc Chimera Protein

Product Code. 18725726
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18725726

Brand: R&D Systems™ 10804S3025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by the ability to inhibit the proliferation of HUVECumbilical vein endothelial cells. The ED50 for this effect is 100-600 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NS98.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 56920
Molecular Weight (g/mol) M.W.-observed: 105-125 kDa, under reducing conditions, M.W.-theroretical: 111 kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Semaphorin 3 G protein Human Semaphorin 3 G (Gly23-Thr782)(R557S, R558S, R560S, R561S, R774S, R777S) (N-terminus) Accession # Q9NS98.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias (semaphorin) 3 G, FLJ00014, MGC119473, sem2, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, SEMA3G, Semaphorin sem2, semaphorin-3 G
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein SEMA3G
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.