missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sentrin-specific protease 8 Protein

A DNA sequence encoding the Homo sapiens (Human) Sentrin-specific protease 8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7846-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031141

Quantity Price
1 € 1180.00 / 500µg
3 € 1100.47 / 500µg
Estimated Shipment
28-10-2020
Add to basket


Specifications

Specifications

NP_001159812.1
Sentrin-specific protease 8 Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
123228
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SENP8
Recombinant Protein
E. coli
MDPVVLSYMD SLLRQSDVSL LDPPSWLNDH IIGFAFEYFA NSQFHDCSDH VSFISPEVTQ FIKCTSNPAE IAMFLEPLDL PNKRVVFLAI NDNSNQAAGG THWSLLVYLQ DKNSFFHYDS HSRSNSVHAK QVAEKLEAFL GRKGDKLAFV EEKAPAQQNS YDCGMYVICN TEALCQNFFR QQTESLLQLL TPAYITKKRG EWKDLITTLA KK
Videos
Documents